2,4-Dihydroxypyrido[3,2-d]pyrimidine 95%
Supplier: Apollo Scientific
OR965167-1G OR965167-5G
APOSOR965167-1GEA 232 EUR
APOSOR965167-1G APOSOR965167-5G
2,4-Dihydroxypyrido[3,2-d]pyrimidine 95%
2,4-Dihydroxypyrido[3,2-d]pyrimidine
Formula: C₇H₅N₃O₂
MW: 163.134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18070985
CAS Number: 37538-68-4