3-Cyclopropyl-3-oxopropanenitrile 95%
Supplier: Apollo Scientific
Warning
OR965365-25G
APOSOR965365-25GEA 169 EUR
APOSOR965365-25G
3-Cyclopropyl-3-oxopropanenitrile 95%
3-Cyclopropyl-3-oxopropanenitrile
Formula: C₆H₇NO
MW: 109.126 g/mol
Boiling Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD04114392
CAS Number: 118431-88-2