3-Bromo-2,3-dihydrothiophene 1,1-dioxide 96%
Supplier: Apollo Scientific
OR965428-1G OR965428-5G
APOSOR965428-1GEA 290 EUR
APOSOR965428-1G APOSOR965428-5G
3-Bromo-2,3-dihydrothiophene 1,1-dioxide 96%
3-Bromo-2,3-dihydrothiophene 1,1-dioxide
Formula: C₄H₅BrO₂S
MW: 197.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00462449
CAS Number: 53336-42-8