5-Nitro-1,3-benzoxazole-2-thiol 95%
Supplier: Apollo Scientific
Warning
OR965833-10G OR965833-1G OR965833-5G
APOSOR965833-10GEA 884 EUR
APOSOR965833-10G APOSOR965833-1G APOSOR965833-5G
5-Nitro-1,3-benzoxazole-2-thiol 95%
5-Nitro-1,3-benzoxazole-2-thiol
Formula: C₇H₄N₂O₃S
MW: 196.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00524789
CAS Number: 22876-21-7