(1R)-1-[2-(Trifluoromethyl)phenyl]ethan-1-ol 97%
Supplier: Apollo Scientific
Warning
Synonyms: (R)-1-(2-(Trifluoromethyl)phenyl)ethanol
PC0586-1G PC0586-250MG
APOSPC0586-1GEA 499 EUR
APOSPC0586-1G APOSPC0586-250MG
(1R)-1-[2-(Trifluoromethyl)phenyl]ethan-1-ol 97%
(1R)-1-[2-(Trifluoromethyl)phenyl]ethan-1-ol
Formula: C₉H₉F₃O
MW: 190.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093000
CAS Number: 127852-29-3