(1S)-1-[2-(Trifluoromethyl)phenyl]ethan-1-ol
Supplier: Apollo Scientific
Warning
Synonyms: S-OTF-PEL, (S)-1-(2-(Trifluoromethyl)phenyl)ethanol
PC0587-1G PC0587-250MG PC0587-5G
APOSPC0587-1GEA 52.2 EUR
APOSPC0587-1G APOSPC0587-250MG APOSPC0587-5G
(1S)-1-[2-(Trifluoromethyl)phenyl]ethan-1-ol
(1S)-1-[2-(Trifluoromethyl)phenyl]ethan-1-ol
Formula: C₉H₉F₃O
MW: 190.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093001
CAS Number: 127852-27-1