(1R)-1-[3-(Trifluoromethyl)phenyl]ethan-1-ol 97%
Supplier: Apollo Scientific
Warning
PC0588-1G PC0588-250MG
APOSPC0588-1GEA 1320 EUR
APOSPC0588-1G APOSPC0588-250MG
(1R)-1-[3-(Trifluoromethyl)phenyl]ethan-1-ol 97%
(1R)-1-[3-(Trifluoromethyl)phenyl]ethan-1-ol
Formula: C₉H₉F₃O
MW: 190.162 g/mol
Boiling Pt: 100…102 °C
Density: 1.249 g/cm³ (25 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD02684188
CAS Number: 127852-24-8