(1S)-1-[4-(Trifluoromethyl)phenyl]ethan-1-ol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-[(1S)-1-Hydroxyethyl]benzotrifluoride, (S)-1-(4-(Trifluoromethyl)phenyl)ethanol, (R)-α-Methyl-4-(trifluoromethyl)benzyl alcohol
PC0595-1G PC0595-250MG
APOSPC0595-1GEA 166 EUR
APOSPC0595-1G APOSPC0595-250MG
(1S)-1-[4-(Trifluoromethyl)phenyl]ethan-1-ol 98%
(1S)-1-[4-(Trifluoromethyl)phenyl]ethan-1-ol
Formula: C₉H₉F₃O
MW: 190.162 g/mol
Density: 1.1 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03093008
CAS Number: 99493-93-3