(1R)-1-[2-(Trifluoromethyl)phenyl]ethylamine
Supplier: Apollo Scientific
Warning
Synonyms: (R)-1-(2-(Trifluoromethyl)phenyl)ethanamine, (R)-α-Methyl-2-(trifluoromethyl)benzylamine, 2-[(1R)-1-(Aminoethyl)]benzotrifluoride
PC0597-100MG PC0597-250MG
APOSPC0597-100MGEA 36.4 EUR
APOSPC0597-100MG APOSPC0597-250MG
(1R)-1-[2-(Trifluoromethyl)phenyl]ethylamine
(1R)-1-[2-(Trifluoromethyl)phenyl]ethylamine
Formula: C₉H₁₀F₃N
MW: 189.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093002
CAS Number: 127733-46-4