(1S)-1-[4-(Trifluoromethyl)phenyl]ethylamine 98%
Supplier: Apollo Scientific
Warning
Synonyms: (S)-α-Methyl-4-(trifluoromethyl)benzylamine
PC0601-1G PC0601-250MG
APOSPC0601-1GEA 212 EUR
APOSPC0601-1G APOSPC0601-250MG
(1S)-1-[4-(Trifluoromethyl)phenyl]ethylamine 98%
(1S)-1-[4-(Trifluoromethyl)phenyl]ethylamine
Formula: C₉H₁₀F₃N
MW: 189.178 g/mol
Boiling Pt: 76 °C (20 mm Hg)
Density: 1.03 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03093009
CAS Number: 84499-73-0