Hexakis(1H,1H,7H-perfluoroheptoxy)phosphazene 97%
Supplier: Apollo Scientific
Warning
PC0657-1G PC0657-250MG
APOSPC0657-1GEA 405 EUR
APOSPC0657-1G APOSPC0657-250MG
Hexakis(1H,1H,7H-perfluoroheptoxy)phosphazene 97%
Hexakis(1H,1H,7H-perfluoroheptoxy)phosphazene
Formula: C₄₂H₁₈F₇₂N₃O₆P₃
MW: 2121.415 g/mol
Melting Pt: 47…51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00798143
CAS Number: 3830-74-8