1-Bromo-1,1,2,2-tetrafluorooctane
Supplier: Apollo Scientific
Warning
PC0766-1G PC0766-5G
APOSPC0766-1GEA 21.9 EUR
APOSPC0766-1G APOSPC0766-5G
1-Bromo-1,1,2,2-tetrafluorooctane
1-Bromo-1,1,2,2-tetrafluorooctane
Formula: C₈H₁₃BrF₄
MW: 265.086 g/mol
Boiling Pt: 67…69 °C (20 mm Hg)
Density: 1.7 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00153714
CAS Number: 231630-92-5