2,4-Dinitro-4'-fluorodiphenyl ether
Supplier: Apollo Scientific
Warning
Synonyms: 2,4-Dinitro-1-(4-fluorophenoxy)benzene
PC0814-1G PC0814-5G
APOSPC0814-1GEA 36.4 EUR
APOSPC0814-1G APOSPC0814-5G
2,4-Dinitro-4'-fluorodiphenyl ether
2,4-Dinitro-4'-fluorodiphenyl ether
Formula: C₁₂H₇FN₂O₅
MW: 278.193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01046749
CAS Number: 1033-02-9