1,3-Bis(4-fluorophenyl)-2,3-dibromopropan-1-one
Supplier: Apollo Scientific
Warning
Synonyms: 2,3-Dibromo-4'-fluoro-3-(4-fluorophenyl)propiophenone
PC0818-1G PC0818-5G
APOSPC0818-1GEA 174 EUR
APOSPC0818-1G APOSPC0818-5G
1,3-Bis(4-fluorophenyl)-2,3-dibromopropan-1-one
1,3-Bis(4-fluorophenyl)-2,3-dibromopropan-1-one
Formula: C₁₅H₁₀Br₂F₂O
MW: 404.044 g/mol
Melting Pt: 165…167 °C
Storage Temperature: Ambient
MDL Number: MFCD03788525
CAS Number: 497959-30-5