4-Bromo-2-chloro-5-fluorotoluene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-2-chlor-6-fluorotoluene, 1-Bromo-5-chloro-2-fluoro-4-methylbenzene
PC0864-1G PC0864-5G PC0864-10G
APOSPC0864-1GEA 21.9 EUR
APOSPC0864-1G APOSPC0864-5G APOSPC0864-10G
4-Bromo-2-chloro-5-fluorotoluene 99%
4-Bromo-2-chloro-5-fluorotoluene
Formula: C₇H₅BrClF
MW: 223.47 g/mol
Boiling Pt: 100…103 °C (22 mm Hg)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00236592
CAS Number: 93765-83-4