(S)-2-Amino-1,1,1-trifluoropropane hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: (S)-1,1,1-Trifluoropropan-2-amine hydrochloride
PC0908-1G PC0908-250MG PC0908-5G
APOSPC0908-1GEA 147 EUR
APOSPC0908-1G APOSPC0908-250MG APOSPC0908-5G
(S)-2-Amino-1,1,1-trifluoropropane hydrochloride
(S)-2-Amino-1,1,1-trifluoropropane hydrochloride
Formula: C₃H₆F₃N·ClH
MW: 149.543 g/mol
Melting Pt: 217…221 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD07779094
CAS Number: 125353-44-8