2-Bromo-2,2-difluoroethylamine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-1-bromo-1,1-difluoroethane hydrochloride
PC0973-1G PC0973-250MG
APOSPC0973-1GEA 568 EUR
APOSPC0973-1G APOSPC0973-250MG
2-Bromo-2,2-difluoroethylamine hydrochloride
2-Bromo-2,2-difluoroethylamine hydrochloride
Formula: C₂H₄BrF₂N·ClH
MW: 196.422 g/mol
Melting Pt: 201 °C
Storage Temperature: Ambient
MDL Number: MFCD07784274
CAS Number: 1211120-17-0