3,5-Dimethyl-2,4,6-trifluorobenzoic acid 97%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Carboxy-2,4,6-trifluoro-m-xylene
PC10082-10G PC10082-1G PC10082-5G
APOSPC10082-10GEA 1160 EUR
APOSPC10082-10G APOSPC10082-1G APOSPC10082-5G
3,5-Dimethyl-2,4,6-trifluorobenzoic acid 97%
3,5-Dimethyl-2,4,6-trifluorobenzoic acid
Formula: C₉H₇F₃O₂
MW: 204.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039199
CAS Number: 886762-23-8