Methyl 4-[2,6-dinitro-4-(trifluoromethyl)phenoxy]benzenecarboxylate
Supplier: Apollo Scientific
PC10113-1G PC10113-500MG PC10113-5G
APOSPC10113-1GEA 302 EUR
APOSPC10113-1G APOSPC10113-500MG APOSPC10113-5G
Methyl 4-[2,6-dinitro-4-(trifluoromethyl)phenoxy]benzenecarboxylate
Methyl 4-[2,6-dinitro-4-(trifluoromethyl)phenoxy]benzenecarboxylate
Formula: C₁₅H₉F₃N₂O₇
MW: 386.236 g/mol
Melting Pt: 127…129 °C
Storage Temperature: Ambient
MDL Number: MFCD01568619
CAS Number: 341967-63-3