1,1,1-Trifluoropent-4-en-2-ol
Supplier: Apollo Scientific
Warning
Synonyms: 4-Hydroxy-5,5,5-trifluoropent-1-ene, 5,5,5-Trifluoropent-1-en-4-ol
PC1016-1G PC1016-5G
APOSPC1016-1GEA 219 EUR
APOSPC1016-1G APOSPC1016-5G
1,1,1-Trifluoropent-4-en-2-ol
1,1,1-Trifluoropent-4-en-2-ol
Formula: C₅H₇F₃O
MW: 140.104 g/mol
Boiling Pt: 100…101 °C
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD07784256
CAS Number: 77342-37-1
UN: 1993
ADR: 3,III