2-Chloro-3-fluoroprop-1-ene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloroallyl fluoride, 2-Chloroprop-2-en-1-yl fluoride
PC1025-1G PC1025-5G
APOSPC1025-1GEA 133 EUR
APOSPC1025-1G APOSPC1025-5G
2-Chloro-3-fluoroprop-1-ene 97%
2-Chloro-3-fluoroprop-1-ene
Formula: C₃H₄ClF
MW: 94.515 g/mol
Boiling Pt: 43…44 °C
Density: 1.1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD07784208
CAS Number: 32804-07-2
UN: 1993
ADR: 3,III