N,N-Diethylammonium trifluoromethanesulphonate 60% in aqueous solution
Supplier: Apollo Scientific
Warning
PC10403-1KG PC10403-250G PC10403-25G
APOSPC10403-1KGEA 444 EUR
APOSPC10403-1KG APOSPC10403-250G APOSPC10403-25G
N,N-Diethylammonium trifluoromethanesulphonate 60% in aqueous solution
N,N-Diethylammonium trifluoromethanesulphonate
Formula: CHF₃O₂S·(C₂H₅)₂NH
MW: 207.214 g/mol
Boiling Pt: 98 °C
Density: 1.2 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01940771
CAS Number: 60933-18-8
EINECS: 262-529-5