2-Bromo-4,5-difluorobenzoyl chloride
Supplier: Apollo Scientific
Danger
PC10442-1G PC10442-5G PC10442-10G
APOSPC10442-1GEA 119 EUR
APOSPC10442-1G APOSPC10442-5G APOSPC10442-10G
2-Bromo-4,5-difluorobenzoyl chloride
2-Bromo-4,5-difluorobenzoyl chloride
Formula: C₇H₂BrClF₂O
MW: 255.444 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368698
CAS Number: 103994-89-4
UN: 3265
ADR: 8 III