N-{[4-Bromo-2-(trifluoromethyl)phenyl]carbamothioyl}benzamide 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Benzoyl-3-[4-bromo-2-(trifluoromethyl)phenyl]thiourea, 2-[(Benzoylcarbamothioyl)amino]-5-bromobenzotrifluoride
PC10458-1G PC10458-5G
APOSPC10458-1GEA 52.2 EUR
APOSPC10458-1G APOSPC10458-5G
N-{[4-Bromo-2-(trifluoromethyl)phenyl]carbamothioyl}benzamide 97%
N-{[4-Bromo-2-(trifluoromethyl)phenyl]carbamothioyl}benzamide
Formula: C₁₅H₁₀BrF₃N₂OS
MW: 403.217 g/mol
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD07368704
CAS Number: 887266-94-6