3,5-Difluorophenylthiourea
Supplier: Apollo Scientific
Danger
Synonyms: 1-(3,5-Difluorophenyl)thiourea, 1,3-Difluoro-5-thioureidobenzene
PC10576-1G PC10576-5G
APOSPC10576-1GEA 105 EUR
APOSPC10576-1G APOSPC10576-5G
3,5-Difluorophenylthiourea
3,5-Difluorophenylthiourea
Formula: C₇H₆F₂N₂S
MW: 188.198 g/mol
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD07368707
CAS Number: 791594-33-7
UN: 2811
ADR: 6.1,III