2-Hydroxy-4-(trifluoromethyl)benzonitrile
Supplier: Apollo Scientific
Warning
PC10674-1G PC10674-250MG PC10674-5G
APOSPC10674-1GEA 314 EUR
APOSPC10674-1G APOSPC10674-250MG APOSPC10674-5G
2-Hydroxy-4-(trifluoromethyl)benzonitrile
Formula: C₈H₄F₃NO
Melting Pt: 118…119 °C
Storage Temperature: Ambient
MDL Number: MFCD13186735
CAS Number: 81465-88-5