2-Iodo-4-(trifluoromethyl)benzaldehyde 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Iodo-α,α,α-trifluoro-p-tolualdehyde, 4-Formyl-3-iodobenzotrifluoride
PC10680-1G PC10680-5G PC10680-10G
APOSPC10680-1GEA 55.1 EUR
APOSPC10680-1G APOSPC10680-5G APOSPC10680-10G
2-Iodo-4-(trifluoromethyl)benzaldehyde 97%
2-Iodo-4-(trifluoromethyl)benzaldehyde
Formula: C₈H₄F₃IO
MW: 300.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18398121
CAS Number: 873006-01-0