2-Bromo-5-(trifluoromethoxy)benzyl alcohol 95%
Supplier: Apollo Scientific
Synonyms: 4-Bromo-3-(hydroxymethyl)-α,α,α-trifluoroanisole, 4-Bromo-3-(hydroxymethyl) trifluoromethyl ether, [2-Bromo-5-(trifluoromethoxy)phenyl]methanol
PC1069-1G PC1069-5G
APOSPC1069-1GEA 150 EUR
APOSPC1069-1G APOSPC1069-5G
2-Bromo-5-(trifluoromethoxy)benzyl alcohol 95%
2-Bromo-5-(trifluoromethoxy)benzyl alcohol
Formula: C₈H₆BrF₃O₂
MW: 271.031 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368723
CAS Number: 886763-15-1