1,4-Difluoro-2-iodo-3-(trifluoromethoxy)benzene 95%
Supplier: Apollo Scientific
Warning
PC10739-1G PC10739-250MG PC10739-5G
APOSPC10739-1GEA 261 EUR
APOSPC10739-1G APOSPC10739-250MG APOSPC10739-5G
1,4-Difluoro-2-iodo-3-(trifluoromethoxy)benzene 95%
1,4-Difluoro-2-iodo-3-(trifluoromethoxy)benzene
Formula: C₇H₂F₅IO
MW: 323.987 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32066739
CAS Number: 2366994-14-9