2-Amino-4-(2-fluorophenylamino)-1,3,5-triazine 97%
Supplier: Apollo Scientific
Warning
PC1076R-100MG PC1076R-1G
APOSPC1076R-100MGEA 29 EUR
APOSPC1076R-100MG APOSPC1076R-1G
2-Amino-4-(2-fluorophenylamino)-1,3,5-triazine 97%
2-Amino-4-(2-fluorophenylamino)-1,3,5-triazine
Formula: C₉H₈FN₅
Melting Pt: 201…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00052347
CAS Number: 66088-45-7