2-Amino-3-(methylthiomethyl)benzotrifluoride 96%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Methylthiomethyl)-6-(trifluoromethyl)aniline
PC1079F-1G PC1079F-5G
APOSPC1079F-1GEA 92.8 EUR
APOSPC1079F-1G APOSPC1079F-5G
2-Amino-3-(methylthiomethyl)benzotrifluoride 96%
2-Amino-3-(methylthiomethyl)benzotrifluoride
Formula: C₉H₁₀F₃NS
MW: 221.243 g/mol
Boiling Pt: 71…73 °C (6 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00052842
CAS Number: 88301-96-6