2-Bromo-5-(trifluoromethoxy)benzoic acid 99%
Supplier: Apollo Scientific
Warning
PC1082-1G PC1082-5G PC1082-25G
APOSPC1082-1GEA 108 EUR
APOSPC1082-1G APOSPC1082-5G APOSPC1082-25G
2-Bromo-5-(trifluoromethoxy)benzoic acid 99%
2-Bromo-5-(trifluoromethoxy)benzoic acid
Formula: C₈H₄BrF₃O₃
MW: 285.015 g/mol
Melting Pt: 77…78 °C
Storage Temperature: Ambient
MDL Number: MFCD07368726
CAS Number: 403646-48-0