2-Fluoro-5-(trifluoromethoxy)toluene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Fluoro-3-methylphenyl trifluoromethyl ether, 1-Fluoro-2-methyl-4-(trifluoromethoxy)benzene
PC1091-1G PC1091-5G
APOSPC1091-1GEA 92.8 EUR
APOSPC1091-1G APOSPC1091-5G
2-Fluoro-5-(trifluoromethoxy)toluene 98%
2-Fluoro-5-(trifluoromethoxy)toluene
Formula: C₈H₆F₄O
MW: 194.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368733
CAS Number: 86256-57-7