8-Fluoro-7-(trifluoromethyl)quinolin-4-ol
Supplier: Apollo Scientific
Warning
PC110011-1G PC110011-250MG
APOSPC110011-1GEA 241 EUR
APOSPC110011-1G APOSPC110011-250MG
8-Fluoro-7-(trifluoromethyl)quinolin-4-ol
8-Fluoro-7-(trifluoromethyl)quinolin-4-ol
Formula: C₁₀H₅F₄NO
MW: 231.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32200323
CAS Number: 693818-12-1