3-Amino-2-(trifluoromethyl)benzamide
Supplier: Apollo Scientific
Warning
PC110023-1G PC110023-250MG
APOSPC110023-1GEA 433 EUR
APOSPC110023-1G APOSPC110023-250MG
3-Amino-2-(trifluoromethyl)benzamide
3-Amino-2-(trifluoromethyl)benzamide
Formula: C₈H₇F₃N₂O
MW: 204.149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29097134
CAS Number: 1369923-89-6