3-(3,5-Difluorophenyl)-1H-pyrazole
Supplier: Apollo Scientific
PC110031-1G PC110031-250MG
APOSPC110031-1GEA 219 EUR
APOSPC110031-1G APOSPC110031-250MG
3-(3,5-Difluorophenyl)-1H-pyrazole
3-(3,5-Difluorophenyl)-1H-pyrazole
Formula: C₉H₆F₂N₂
MW: 180.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05664754
CAS Number: 154258-58-9