3-Amino-4,5-difluorobenzonitrile
Supplier: Apollo Scientific
PC110112-1G
PC110112-5G
APOSPC110112-1GEA
435
EUR
APOSPC110112-1G
APOSPC110112-5G
3-Amino-4,5-difluorobenzonitrile
3-Amino-4,5-difluorobenzonitrile
Formula:
C₇H₄F₂N₂ MW: 154.117 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22490583 CAS Number: 1805635-16-8 |