4'-Fluoro-[1,1'-biphenyl]-2-carboxylic acid
Supplier: Apollo Scientific
Warning
PC11142-250MG PC11142-1G
APOSPC11142-250MGEA 33.5 EUR
APOSPC11142-250MG APOSPC11142-1G
4'-Fluoro-[1,1'-biphenyl]-2-carboxylic acid
4'-Fluoro-[1,1'-biphenyl]-2-carboxylic acid
Formula: C₁₃H₉FO₂
MW: 216.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03002065
CAS Number: 1841-57-2