2'-(Trifluoromethyl)-[1,1'-biphenyl]-3-amine hydrochloride
Supplier: Apollo Scientific
Warning
PC11162-1G PC11162-250MG
APOSPC11162-1GEA 466 EUR
APOSPC11162-1G APOSPC11162-250MG
2'-(Trifluoromethyl)-[1,1'-biphenyl]-3-amine hydrochloride
2'-(Trifluoromethyl)-[1,1'-biphenyl]-3-amine hydrochloride
Formula: C₁₃H₁₀F₃N·ClH
MW: 273.681 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06739425
CAS Number: 811842-39-4