3,6-Difluoro-2-methoxybenzoic acid
Supplier: Apollo Scientific
Warning
Synonyms: 3,6-Difluoro-o-anisic acid, 2-Carboxy-3,6-difluoroanisole
PC1138-1G PC1138-5G
APOSPC1138-1GEA 43.5 EUR
APOSPC1138-1G APOSPC1138-5G
3,6-Difluoro-2-methoxybenzoic acid
3,6-Difluoro-2-methoxybenzoic acid
Formula: C₈H₆F₂O₃
MW: 188.128 g/mol
Melting Pt: 85…86 °C
Storage Temperature: Ambient
MDL Number: MFCD08275290
CAS Number: 887267-03-0