3'-Fluoro[1,1'-biphenyl]-4-carboxylic acid 98%
Supplier: Apollo Scientific
PC1141-25G PC1141-5G
APOSPC1141-25GEA 510 EUR
APOSPC1141-25G APOSPC1141-5G
3'-Fluoro[1,1'-biphenyl]-4-carboxylic acid 98%
3'-Fluoro[1,1'-biphenyl]-4-carboxylic acid
Formula: C₁₃H₉FO₂
MW: 216.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03424609
CAS Number: 1841-58-3