2-Nitro-4-(trifluoromethyl)phenylhydrazine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: 4-Hydrazino-3-nitrobenzotrifluoride hydrochloride, 2-Hydrazino-5-(trifluoromethyl)nitrobenzene hydrochloride
PC1277-1G PC1277-25G PC1277-5G
APOSPC1277-1GEA 140 EUR
APOSPC1277-1G APOSPC1277-25G APOSPC1277-5G
2-Nitro-4-(trifluoromethyl)phenylhydrazine hydrochloride
2-Nitro-4-(trifluoromethyl)phenylhydrazine hydrochloride
Formula: C₇H₆F₃N₃O₂·ClH
MW: 257.598 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04966802
CAS Number: 1049748-69-7