4-Methyl-2,3,5,6-tetrafluoropyridine 99+%
Supplier: Apollo Scientific
Warning
PC1393-1G PC1393-250MG PC1393-5G
APOSPC1393-1GEA 232 EUR
APOSPC1393-1G APOSPC1393-250MG APOSPC1393-5G
4-Methyl-2,3,5,6-tetrafluoropyridine 99+%
4-Methyl-2,3,5,6-tetrafluoropyridine
Formula: C₆H₃F₄N
MW: 165.088 g/mol
Boiling Pt: 130…131 °C
Density: 1.44 g/cm³ (25 °C)
Flash Pt: 32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012345
CAS Number: 16297-14-6
UN: 1993
ADR: 3,III