1-[2-(Trifluoromethoxy)benzyl]piperazine
Supplier: Apollo Scientific
Warning
Synonyms: 1-[(Piperazin-1-yl)methyl]-2-(trifluoromethoxy)benzene
PC1411-1G PC1411-25G PC1411-5G
APOSPC1411-1GEA 111 EUR
APOSPC1411-1G APOSPC1411-25G APOSPC1411-5G
1-[2-(Trifluoromethoxy)benzyl]piperazine
1-[2-(Trifluoromethoxy)benzyl]piperazine
Formula: C₁₂H₁₅F₃N₂O
MW: 260.256 g/mol
Boiling Pt: 78…80 °C (0.02 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD07368743
CAS Number: 886762-54-5