3,4-Dichloro-2-fluoroaniline 98%
Supplier: Apollo Scientific
|
Warning
|
PC14291-100G
PC14291-25G
PC14291-50G
APOSPC14291-100GEA
185
EUR
APOSPC14291-100G
APOSPC14291-25G
APOSPC14291-50G
3,4-Dichloro-2-fluoroaniline 98%
3,4-Dichloro-2-fluoroaniline
Formula:
C₆H₄Cl₂FN Melting Pt: 53…56 °C Density: 1.5 g/cm³ (25 °C) Flash Pt: 109 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039241 CAS Number: 886762-39-6 UN: 2811 ADR: 6.1,III |