2-Methoxy-4-(trifluoromethoxy)bromobenzene 96%
Supplier: Apollo Scientific
Synonyms: 1-Bromo-2-methoxy-4-(trifluoromethoxy)benzene
PC1574-10G PC1574-1G PC1574-5G PC1574-25G
APOSPC1574-10GEA 492 EUR
APOSPC1574-10G APOSPC1574-1G APOSPC1574-5G APOSPC1574-25G
2-Methoxy-4-(trifluoromethoxy)bromobenzene 96%
2-Methoxy-4-(trifluoromethoxy)bromobenzene
Formula: C₈H₆BrF₃O₂
MW: 271.031 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458014
CAS Number: 848360-85-0