3-Bromo-1,1,1-trifluorobutan-2-ol 95%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-2-hydroxy-1,1,1-trifluorobutane
PC1583C-1G PC1583C-250MG
APOSPC1583C-1GEA 364 EUR
APOSPC1583C-1G APOSPC1583C-250MG
3-Bromo-1,1,1-trifluorobutan-2-ol 95%
3-Bromo-1,1,1-trifluorobutan-2-ol
Formula: C₄H₆BrF₃O
MW: 206.989 g/mol
Boiling Pt: 130…132 °C
Density: 1.6771 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236598
CAS Number: 382-00-3
UN: 1993
ADR: 3,III