3-Bromo-1,1,1-trifluorobutan-2-one 95%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-2-oxo-1,1,1-trifluorobutane, 3-Bromo-3-methyl-1,1,1-trifluoroacetone
PC1583E-5G PC1583E-1G
APOSPC1583E-5GEA 379 EUR
APOSPC1583E-5G APOSPC1583E-1G
3-Bromo-1,1,1-trifluorobutan-2-one 95%
3-Bromo-1,1,1-trifluorobutan-2-one
Formula: C₄H₄BrF₃O
MW: 204.973 g/mol
Boiling Pt: 93 °C (748 mm Hg)
Density: 1.64 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00236599
CAS Number: 382-01-4
UN: 1993
ADR: 3,III