n-Butyl 1,1,2,2-tetrafluoroethyl ether 97%
Supplier: Apollo Scientific
Warning
PC1612-25G PC1612-5G
APOSPC1612-25GEA 50.9 EUR
APOSPC1612-25G APOSPC1612-5G
n-Butyl 1,1,2,2-tetrafluoroethyl ether 97%
n-Butyl 1,1,2,2-tetrafluoroethyl ether
Formula: C₆H₁₀F₄O
MW: 174.137 g/mol
Boiling Pt: 49 °C (113 mm Hg)
Density: 1.1147 g/cm³ (25 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00042107
CAS Number: 358-37-2
UN: 3271
ADR: 3,II