2-Chloro-2,2-difluoroethane-1,1-diol 95%
Supplier: Apollo Scientific
Warning
PC1728-1G PC1728-5G
APOSPC1728-1GEA 87 EUR
APOSPC1728-1G APOSPC1728-5G
2-Chloro-2,2-difluoroethane-1,1-diol 95%
2-Chloro-2,2-difluoroethane-1,1-diol
Formula: C₂H₃ClF₂O₂
MW: 132.494 g/mol
Density: 1.384 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042464
CAS Number: 63034-47-9
UN: 2810
ADR: 6.1 III